5-Bromo-2-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-2-(trifluoromethyl)phenyl)methanol, [5-Bromo-2-(trifluoromethyl)phenyl]methanol

PC500758-1G PC500758-250MG
APOSPC500758-1GEA 393 EUR
APOSPC500758-1G APOSPC500758-250MG
5-Bromo-2-(trifluoromethyl)benzyl alcohol
5-Bromo-2-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆BrF₃O
Molecular Weight: 255.032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185398
CAS-Nummer: 1214349-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR