5-Bromo-2-(trifluoromethyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC500760-1G PC500760-250MG PC500760-100MG
APOSPC500760-1GEA 219 EUR
APOSPC500760-1G APOSPC500760-250MG APOSPC500760-100MG
5-Bromo-2-(trifluoromethyl)benzyl bromide
5-Bromo-2-(trifluoromethyl)benzyl bromide
Formel: C₈H₅Br₂F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13185415
CAS-Nummer: 1214327-32-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR