Methyl 5-bromo-2-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific

PC500761-1G PC500761-250MG PC500761-5G
APOSPC500761-1GEA 0 EUR
APOSPC500761-1G APOSPC500761-250MG APOSPC500761-5G
Methyl 5-bromo-2-(trifluoromethyl)benzoate
Methyl 5-bromo-2-(trifluoromethyl)benzoate
Formel: C₉H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14697999
CAS-Nummer: 842136-32-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR