6-Fluorophthalide 95+%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoroisobenzofuran-1(3H)-one

PC500778-250MG
APOSPC500778-250MGEA 183 EUR
APOSPC500778-250MG
6-Fluorophthalide 95+%
6-Fluorophthalide
Formel: C₈H₅FO₂
Molecular Weight: 152,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053086
CAS-Nummer: 23932-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR