2-Bromo-3-fluoroisonicotinaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3-fluoro-4-formylpyridine, 2-Bromo-3-fluoropyridine-4-carboxaldehyde

PC50078-1G
APOSPC50078-1GEA 48 EUR
APOSPC50078-1G
2-Bromo-3-fluoroisonicotinaldehyde
2-Bromo-3-fluoroisonicotinaldehyde
Formel: C₆H₃BrFNO
Molecular Weight: 203.997 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16606504
CAS-Nummer: 1227572-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR