5-Fluoro-2-methoxynicotinaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-methoxypyridine-3-carboxaldehyde

PC50081-1G
APOSPC50081-1GEA 21.9 EUR
APOSPC50081-1G
5-Fluoro-2-methoxynicotinaldehyde
5-Fluoro-2-methoxynicotinaldehyde
Formel: C₇H₆FNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD04972398
CAS-Nummer: 351410-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR