2-Bromo-6-(trifluoromethyl)nicotinaldehyde 85+%

Lieferant: Apollo Scientific
Achtung

PC50083-1G
APOSPC50083-1GEA 69.6 EUR
APOSPC50083-1G
2-Bromo-6-(trifluoromethyl)nicotinaldehyde 85+%
2-Bromo-6-(trifluoromethyl)nicotinaldehyde
Formel: C₇H₃BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13193090
CAS-Nummer: 1192263-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR