1-Hexyl-3-methylimidazoliumhexafluorophosphat 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

PC50085-100G
APOSPC50085-100GEA 125 EUR
APOSPC50085-100G
1-Hexyl-3-methylimidazoliumhexafluorophosphat 95+%
1-Hexyl-3-methylimidazolium hexafluorophosphate
Formel: C₁₀H₁₉F₆N₂P
Molecular Weight: 312,24 g/mol
Dichte: 1,419 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD03093296
CAS-Nummer: 304680-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR