4-(2,2,2-Trifluoroethoxy)benzoic acid 95+%

Lieferant: Apollo Scientific

PC500851-5G
APOSPC500851-5GEA 137 EUR
APOSPC500851-5G
4-(2,2,2-Trifluoroethoxy)benzoic acid 95+%
4-(2,2,2-Trifluoroethoxy)benzoic acid
Formel: C₉H₇F₃O₃
Molecular Weight: 220,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05669500
CAS-Nummer: 27914-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR