2-Fluor-5-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

PC5009-25G PC5009-5G PC5009-1G
APOSPC5009-25GEA 168 EUR
APOSPC5009-25G APOSPC5009-5G APOSPC5009-1G
2-Fluor-5-nitrophenol 98%
2-Fluor-5-nitrophenol
Formel: C₆H₄FNO₃
Molecular Weight: 157.1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09054718
CAS-Nummer: 22510-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR