2-(2,2-Difluoroethoxy)bromobenzene

Lieferant: Apollo Scientific

PC500912-1G PC500912-5G
APOSPC500912-1GEA 199 EUR
APOSPC500912-1G APOSPC500912-5G
2-(2,2-Difluoroethoxy)bromobenzene
2-(2,2-Difluoroethoxy)bromobenzene
Formel: C₈H₇BrF₂O
Siedepunkt: 133 °C (5 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD11036116
CAS-Nummer: 946605-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR