1-Brom-3-(2,2-difluorethoxy)benzol

Lieferant: Apollo Scientific
Achtung

PC500915-1G PC500915-250MG PC500915-100MG
APOSPC500915-1GEA 0 EUR
APOSPC500915-1G APOSPC500915-250MG APOSPC500915-100MG
1-Brom-3-(2,2-difluorethoxy)benzol
1-Bromo-3-(2,2-difluoroethoxy)benzene
Formel: C₈H₇BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11036117
CAS-Nummer: 958454-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR