3-(2,2-Difluoroethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC500935-1G PC500935-250MG PC500935-5G
APOSPC500935-1GEA 0 EUR
APOSPC500935-1G APOSPC500935-250MG APOSPC500935-5G
3-(2,2-Difluoroethoxy)benzyl bromide 95%
3-(2,2-Difluoroethoxy)benzyl bromide
Formel: C₉H₉BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12768652
CAS-Nummer: 864054-55-7
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR