4-Fluoro-6-(trifluoromethyl)-1H-benzimidazole
Lieferant: Apollo Scientific
Achtung
|
PC500991-1G
APOSPC500991-1GEA
509
EUR
APOSPC500991-1G
4-Fluoro-6-(trifluoromethyl)-1H-benzimidazole
4-Fluoro-6-(trifluoromethyl)-1H-benzimidazole
Formel:
C₈H₄F₄N₂ |
MDL:
MFCD28154923 CAS-Nummer: 1360891-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...