4-Bromo-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole

Lieferant: Apollo Scientific
Gefahr

PC501009-1G PC501009-250MG
APOSPC501009-1GEA 609 EUR
APOSPC501009-1G APOSPC501009-250MG
4-Bromo-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
4-Bromo-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
Formel: C₈H₃BrF₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28154938
CAS-Nummer: 1822861-98-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR