2-Fluoro-5-iodobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC50104-1G
APOSPC50104-1GEA 0 EUR
APOSPC50104-1G
2-Fluoro-5-iodobenzyl bromide
2-Fluoro-5-iodobenzyl bromide
Formel: C₇H₅BrFI
Lagertemperatur: Raumtemperatur
MDL: MFCD18396956
CAS-Nummer: 442910-33-0
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR