3-Chloro-4-fluorobenzene -1,2-diamine 95%
Lieferant: Apollo Scientific
PC501050-1G
PC501050-250MG
APOSPC501050-1GEA
729
EUR
APOSPC501050-1G
APOSPC501050-250MG
3-Chloro-4-fluorobenzene -1,2-diamine 95%
3-Chloro-4-fluorobenzene -1,2-diamine
Formel:
C₆H₆ClFN₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD18804697 CAS-Nummer: 132915-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...