3-Chloro-5-fluorobenzene -1,2-diamine

Lieferant: Apollo Scientific

PC501051-1G PC501051-250MG
APOSPC501051-1GEA 0 EUR
APOSPC501051-1G APOSPC501051-250MG
3-Chloro-5-fluorobenzene -1,2-diamine
3-Chloro-5-fluorobenzene -1,2-diamine
Formel: C₆H₆ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20723094
CAS-Nummer: 153505-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR