5-Bromo-4-fluoro-3-(trifluoromethyl)benzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

PC50108-1G PC50108-250MG
APOSPC50108-1GEA 451 EUR
APOSPC50108-1G APOSPC50108-250MG
5-Bromo-4-fluoro-3-(trifluoromethyl)benzene-1,2-diamine
5-Bromo-4-fluoro-3-(trifluoromethyl)benzene-1,2-diamine
Formel: C₇H₅BrF₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28515424
CAS-Nummer: 1820706-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR