Lithium trifluoroacetate 97%

Lieferant: Apollo Scientific
Achtung

PC5011-100G PC5011-10G PC5011-50G
APOSPC5011-100GEA 221 EUR
APOSPC5011-100G APOSPC5011-10G APOSPC5011-50G
Lithium trifluoroacetate 97%
Lithium trifluoroacetate

Useful for the preparation of trifluoroketones; treatment with BuLi gives 61% yield of 1,1,1-trifluoro-2-hexanone;also reacts with Grignard reagents to give ketones.

Formel: LiCO₂CF₃
Molecular Weight: 119.96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013216
CAS-Nummer: 2923-17-3
EINECS: 220-878-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR