3-Fluoro-2-methyl-4-nitroaniline 98%

Lieferant: Apollo Scientific

PC501106-250MG
APOSPC501106-250MGEA 209 EUR
APOSPC501106-250MG
3-Fluoro-2-methyl-4-nitroaniline 98%
3-Fluoro-2-methyl-4-nitroaniline
Formel: C₇H₇FN₂O₂
Molecular Weight: 170,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026961
CAS-Nummer: 1000342-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR