3-Bromo-5-fluoro-4-iodobenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

PC501118-1G PC501118-250MG PC501118-5G
APOSPC501118-1GEA 1070 EUR
APOSPC501118-1G APOSPC501118-250MG APOSPC501118-5G
3-Bromo-5-fluoro-4-iodobenzotrifluoride 95%
3-Bromo-5-fluoro-4-iodobenzotrifluoride
Formel: C₇H₂BrF₄I
Lagertemperatur: Raumtemperatur
MDL: MFCD28348453
CAS-Nummer: 1934724-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR