4-Bromo-2-fluoro-3-methylbenzoic acid 95+%
Lieferant: Apollo Scientific
PC501158-1G
APOSPC501158-1GEA
105
EUR
APOSPC501158-1G
4-Bromo-2-fluoro-3-methylbenzoic acid 95+%
4-Bromo-2-fluoro-3-methylbenzoic acid
Formel:
C₈H₆BrFO₂ Molecular Weight: 233,034 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22052783 CAS-Nummer: 194804-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...