5-Chloro-4-fluoro-2-nitrophenol

Lieferant: Apollo Scientific

PC501173-5G
APOSPC501173-5GEA 112 EUR
APOSPC501173-5G
5-Chloro-4-fluoro-2-nitrophenol
5-Chloro-4-fluoro-2-nitrophenol
Formel: C6H3ClFNO3
Lagertemperatur: Raumtemperatur
MDL: MFCD18072578
CAS-Nummer: 65001-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR