2-Fluor-3-nitrobenzylalkohol

Lieferant: Apollo Scientific
Achtung

PC501174-1G PC501174-250MG PC501174-5G
APOSPC501174-1GEA 42.2 EUR
APOSPC501174-1G APOSPC501174-250MG APOSPC501174-5G
2-Fluor-3-nitrobenzylalkohol
2-Fluor-3-nitrobenzylalkohol
Formel: C₇H₆FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09835208
CAS-Nummer: 946126-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR