2-Fluor-3-nitrobenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
PC501174-1G
PC501174-250MG
PC501174-5G
APOSPC501174-1GEA
42.2
EUR
APOSPC501174-1G
APOSPC501174-250MG
APOSPC501174-5G
2-Fluor-3-nitrobenzylalkohol
2-Fluor-3-nitrobenzylalkohol
Formel:
C₇H₆FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835208 CAS-Nummer: 946126-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...