2-Fluor-3-nitrobenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC501176-250MG
APOSPC501176-250MGEA
36.4
EUR
APOSPC501176-250MG
2-Fluor-3-nitrobenzaldehyd
2-Fluoro-3-nitrobenzaldehyde
Formel:
C₇H₄FNO₃ Molecular Weight: 169.1100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669885 CAS-Nummer: 96516-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...