6-Bromo-2-fluoro-3-nitrotoluene

Lieferant: Apollo Scientific

PC501226-1G
APOSPC501226-1GEA 512 EUR
APOSPC501226-1G
6-Bromo-2-fluoro-3-nitrotoluene
6-Bromo-2-fluoro-3-nitrotoluene
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27935488
CAS-Nummer: 1806979-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR