2-(2,2-Difluoroethoxy)aniline

Lieferant: Apollo Scientific
Achtung

PC501234-1G PC501234-5G
APOSPC501234-1GEA 0 EUR
APOSPC501234-1G APOSPC501234-5G
2-(2,2-Difluoroethoxy)aniline
2-(2,2-Difluoroethoxy)aniline
Formel: C₈H₉F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08700290
CAS-Nummer: 937606-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR