4-Bromo-2,3-difluoro-6-nitroaniline

Lieferant: Apollo Scientific

PC501256-1G PC501256-5G
APOSPC501256-1GEA 251 EUR
APOSPC501256-1G APOSPC501256-5G
4-Bromo-2,3-difluoro-6-nitroaniline
4-Bromo-2,3-difluoro-6-nitroaniline
Formel: C₆H₃BrF₂N₂O₂
Molecular Weight: 253.001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22375090
CAS-Nummer: 626238-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR