3-(Ethoxycarbonyl)-4-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid

PC5013-1G PC5013-5G
APOSPC5013-1GEA 123 EUR
APOSPC5013-1G APOSPC5013-5G
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure 98%
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211.9827 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08061836
CAS-Nummer: 874219-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR