4-(Trifluoromethoxy)-3-(trifluoromethyl)phenol

Lieferant: Apollo Scientific
Gefahr

PC501341-1G PC501341-250MG
APOSPC501341-1GEA 0 EUR
APOSPC501341-1G APOSPC501341-250MG
4-(Trifluoromethoxy)-3-(trifluoromethyl)phenol
4-(Trifluoromethoxy)-3-(trifluoromethyl)phenol
Formel: C₈H₄F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28348599
CAS-Nummer: 120355-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR