3-Fluoro-4-(methylsulphonyl)benzyl bromide

Lieferant: Apollo Scientific
Achtung

PC501365-1G PC501365-250MG PC501365-5G
APOSPC501365-1GEA 0 EUR
APOSPC501365-1G APOSPC501365-250MG APOSPC501365-5G
3-Fluoro-4-(methylsulphonyl)benzyl bromide
3-Fluoro-4-(methylsulphonyl)benzyl bromide
Formel: C₈H₈BrFO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD20485579
CAS-Nummer: 942199-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR