4-Fluoro-3-(methylsulphonyl)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

PC501366-1G PC501366-250MG PC501366-5G
APOSPC501366-1GEA 696 EUR
APOSPC501366-1G APOSPC501366-250MG APOSPC501366-5G
4-Fluoro-3-(methylsulphonyl)benzyl alcohol 95%
4-Fluoro-3-(methylsulphonyl)benzyl alcohol
Formel: C₈H₉FO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD26051381
CAS-Nummer: 1192347-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR