4-Fluoro-3-(methylsulphonyl)benzyl alcohol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501366-1G
PC501366-250MG
PC501366-5G
APOSPC501366-1GEA
696
EUR
APOSPC501366-1G
APOSPC501366-250MG
APOSPC501366-5G
4-Fluoro-3-(methylsulphonyl)benzyl alcohol 95%
4-Fluoro-3-(methylsulphonyl)benzyl alcohol
Formel:
C₈H₉FO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD26051381 CAS-Nummer: 1192347-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...