2-Fluoro-4-(methylsulphonyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

PC501371-1G PC501371-250MG
APOSPC501371-1GEA 0 EUR
APOSPC501371-1G APOSPC501371-250MG
2-Fluoro-4-(methylsulphonyl)benzonitrile
2-Fluoro-4-(methylsulphonyl)benzonitrile
Formel: C₈H₆FNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD18378490
CAS-Nummer: 411233-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR