2-Fluoro-4-(methylsulphonyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC501375-1G PC501375-250MG PC501375-5G
APOSPC501375-1GEA 0 EUR
APOSPC501375-1G APOSPC501375-250MG APOSPC501375-5G
2-Fluoro-4-(methylsulphonyl)benzyl bromide
2-Fluoro-4-(methylsulphonyl)benzyl bromide
Formel: C₈H₈BrFO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28348619
CAS-Nummer: 1401796-84-6
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR