3-(Ethylcarbamoyl)-5-fluorobenzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Borono-N-ethyl-5-fluorobenzamide

PC5014-1G
APOSPC5014-1GEA 205 EUR
APOSPC5014-1G
3-(Ethylcarbamoyl)-5-fluorobenzeneboronic acid 98%
3-(Ethylcarbamoyl)-5-fluorobenzeneboronic acid
Formel: C₉H₁₁BFNO₃
Molecular Weight: 210,998 g/mol
Schmelzpunkt: 288…290 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235055
CAS-Nummer: 874219-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR