5-Bromo-3-fluoroisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
PC501400-1G
PC501400-5G
APOSPC501400-1GEA
0
EUR
APOSPC501400-1G
APOSPC501400-5G
5-Bromo-3-fluoroisatoic anhydride
5-Bromo-3-fluoroisatoic anhydride
Formel:
C₈H₃BrFNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11899541 CAS-Nummer: 1049118-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...