4-Chloro-3-(trifluoromethoxy)nitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
PC501403-1G
PC501403-250MG
PC501403-5G
APOSPC501403-1GEA
0
EUR
APOSPC501403-1G
APOSPC501403-250MG
APOSPC501403-5G
4-Chloro-3-(trifluoromethoxy)nitrobenzene
4-Chloro-3-(trifluoromethoxy)nitrobenzene
Formel:
C₇H₃ClF₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393864 CAS-Nummer: 113421-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...