4-Chloro-3-(trifluoromethoxy)nitrobenzene

Lieferant: Apollo Scientific
Achtung

PC501403-1G PC501403-250MG PC501403-5G
APOSPC501403-1GEA 0 EUR
APOSPC501403-1G APOSPC501403-250MG APOSPC501403-5G
4-Chloro-3-(trifluoromethoxy)nitrobenzene
4-Chloro-3-(trifluoromethoxy)nitrobenzene
Formel: C₇H₃ClF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18393864
CAS-Nummer: 113421-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR