3-Fluor-5-nitrobenzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

PC501430-5G
APOSPC501430-5GEA 172 EUR
APOSPC501430-5G
3-Fluor-5-nitrobenzaldehyd 97%
3-Fluoro-5-nitrobenzaldehyde
Formel: C₇H₄FNO₃
Molecular Weight: 169.1100 g/mol
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11111000
CAS-Nummer: 108159-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR