Methyl 5-(bromomethyl)-2-fluorobenzoate 95%

Lieferant: Apollo Scientific

PC501442-250MG
APOSPC501442-250MGEA 0 EUR
APOSPC501442-250MG
Methyl 5-(bromomethyl)-2-fluorobenzoate 95%
Methyl 5-(bromomethyl)-2-fluorobenzoate
Formel: C₉H₈BrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD25459290
CAS-Nummer: 709-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR