Methyl 4-(bromomethyl)-2-fluorobenzoate 95%

Lieferant: Apollo Scientific

PC501448-250MG
APOSPC501448-250MGEA 0 EUR
APOSPC501448-250MG
Methyl 4-(bromomethyl)-2-fluorobenzoate 95%
Methyl 4-(bromomethyl)-2-fluorobenzoate
Formel: C₉H₈BrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11976782
CAS-Nummer: 85070-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR