2-Bromophenylsulphur pentafluoride 98%

Lieferant: Apollo Scientific

PC501463-1G PC501463-250MG
APOSPC501463-1GEA 464 EUR
APOSPC501463-1G APOSPC501463-250MG
2-Bromophenylsulphur pentafluoride 98%
2-Bromophenylsulphur pentafluoride
Formel: C₆H₄BrF₅S
Molecular Weight: 283.057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26408179
CAS-Nummer: 1435807-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR