2-Bromophenylsulphur pentafluoride 98%
Lieferant: Apollo Scientific
PC501463-1G
PC501463-250MG
APOSPC501463-1GEA
464
EUR
APOSPC501463-1G
APOSPC501463-250MG
2-Bromophenylsulphur pentafluoride 98%
2-Bromophenylsulphur pentafluoride
Formel:
C₆H₄BrF₅S Molecular Weight: 283.057 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26408179 CAS-Nummer: 1435807-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...