5-Bromo-2-fluoro-4-methylbenzoic acid

Lieferant: Apollo Scientific

PC501554-1G
APOSPC501554-1GEA 238 EUR
APOSPC501554-1G
5-Bromo-2-fluoro-4-methylbenzoic acid
5-Bromo-2-fluoro-4-methylbenzoic acid
Formel: C₈H₆BrFO₂
Molecular Weight: 233,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23712067
CAS-Nummer: 515135-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR