2-Bromo-4,6-difluorobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC501557-1G PC501557-250MG
APOSPC501557-1GEA 203 EUR
APOSPC501557-1G APOSPC501557-250MG
2-Bromo-4,6-difluorobenzyl bromide 95%
2-Bromo-4,6-difluorobenzyl bromide
Formel: C₇H₄Br₂F₂
Molecular Weight: 285.911 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28737919
CAS-Nummer: 1807193-40-3
UN: 3261
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR