5-Fluoro-2-nitrobenzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

PC501559-1G
APOSPC501559-1GEA 0 EUR
APOSPC501559-1G
5-Fluoro-2-nitrobenzenesulphonyl chloride 97%
5-Fluoro-2-nitrobenzenesulphonyl chloride
Formel: C₆H₃ClFNO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD11858121
CAS-Nummer: 82711-97-5
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR