4-Fluor-3-(methylcarbamoyl)phenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid
PC5016-250MG
PC5016-1G
PC5016-5G
APOSPC5016-250MGEA
43.5
EUR
APOSPC5016-250MG
APOSPC5016-1G
APOSPC5016-5G
4-Fluor-3-(methylcarbamoyl)phenylboronsäure 96%
4-Fluor-3-(methylcarbamoyl)phenylboronsäure
Formel:
C₈H₉BFNO₃ Molecular Weight: 196.97 g/mol Schmelzpunkt: 226…228 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235038 CAS-Nummer: 874219-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...